Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146244.4 | NP_666356.2 | 4 | 591 | ||
GenBank mRNA | AK031847.1 | - | 4 | 582 | ||
AK044050.1 | - | 4 | 582 | |||
BC024798.1 | - | 4 | 533 | 5358742 | ||
BC027298.1 | - | 4 | 545 | 3672127 |