Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cchcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146248.2 | NP_666360.2 | 6 | 986 | ||
GenBank mRNA | AK159780.1 | - | 6 | 913 | ||
BC031416.1 | - | 7 | 1075 | 4012768 |