Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146280.1 | NP_666392.1 | 1 | 308 | ||
GenBank mRNA | BC119146.1 | - | 1 | 335 | 8733895 | |
BC120834.1 | - | 1 | 335 | 40129757 |