Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146284.1 | NP_666396.1 | 1 | 670 | ||
GenBank mRNA | BC138405.1 | - | 2064 | 8861425 | ||
BC138406.1 | - | 2064 | 8861426 | |||
BC147436.1 | - | 1 | 726 | 9087435 |