Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000860.5 | NP_000851.2 | 7 | 1202 | ||
NM_001145816.2 | NP_001139288.1 | 6 | 1038 | |||
NM_001256301.1 | NP_001243230.1 | 7 | 932 | |||
NM_001256305.1 | NP_001243234.1 | 5 | 961 | |||
NM_001256306.1 | NP_001243235.1 | 5 | 998 | |||
NM_001256307.1 | NP_001243236.1 | 6 | 1095 | |||
GenBank mRNA | AK058013.1 | - | 7 | 791 | ||
AK296642.1 | - | 7 | 932 | |||
AK300125.1 | - | 5 | 588 | |||
AK300524.1 | - | 6 | 838 | |||
AK300940.1 | - | 6 | 627 | |||
AK314624.1 | - | 7 | 791 | |||
BC018986.2 | - | 7 | 764 | 3638799 | ||
J05594.1 | - | 7 | 772 | |||
L76465.1 | - | 7 | 772 | |||
U63296.1 | - | 5 | 514 | |||
X82460.1 | - | 6 | 591 |