Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146332.1 | NP_666444.1 | 1 | 709 | ||
GenBank mRNA | BC116774.1 | - | 1 | 787 | 40126093 | |
BC116776.1 | - | 1 | 787 | 40126095 |