Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163493.1 | NP_001156965.1 | 14 | 4435 | ||
NM_146258.2 | NP_666370.3 | 14 | 4378 | |||
GenBank mRNA | BC027830.1 | - | 14 | 4390 | 5320966 |