Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr448 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146273.1 | NP_666385.1 | 1 | 899 | ||
GenBank mRNA | BC107336.1 | - | 1 | 891 | ||
BC107337.1 | - | 1 | 899 | 40055003 |