Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000880.3 | NP_000871.1 | 4 | 937 | ||
NM_001199887.1 | NP_001186816.1 | 4 | 937 | |||
XM_011517522.2 | XP_011515824.1 | 4 | 877 | |||
XM_011517523.2 | XP_011515825.1 | 4 | 877 | |||
XM_017013397.1 | XP_016868886.1 | 4 | 821 | |||
GenBank mRNA | AK226000.1 | - | 5 | 1081 | ||
AK291538.1 | - | 4 | 937 | |||
BC047698.1 | - | 4 | 925 | 5748841 | ||
CD013923.1 | - | 4 | 342 | |||
GQ919059.1 | - | 3 | 261 | |||
J04156.1 | - | 4 | 720 |