Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 28 | 4060 | ||
GenBank mRNA | AB208885.1 | - | 25 | 2838 | ||
BC080190.1 | - | 11 | 1405 | |||
X16983.1 | - | 28 | 3339 |