Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321357.1 | NP_001308286.1 | 5, 6 | 1274 | ||
NM_001321358.1 | NP_001308287.1 | 4, 5 | 647 | |||
NM_001321359.1 | NP_001308288.1 | 4, 5 | 642 | |||
NM_001321361.1 | NP_001308290.1 | 4, 5 | 647 | |||
NM_001321362.1 | NP_001308291.1 | 4, 5 | 647 | |||
NM_013392.3 | NP_037524.1 | 4, 5 | 642 | |||
GenBank mRNA | AF113249.1 | - | 4, 5 | 554 | ||
AK001946.1 | - | 4, 5 | 532 | |||
AK027538.1 | - | 4, 5 | 550 | |||
AK122664.1 | - | 5, 6 | 1274 | |||
AK223136.1 | - | 4, 5 | 538 | |||
AK225540.1 | - | 4, 5 | 568 | |||
AK301345.1 | - | 5, 6 | 485 | |||
AL136682.1 | - | 4, 5 | 529 | |||
BC001221.1 | - | 4, 5 | 556 | 3534543 | ||
BQ949042.1 | - | 4, 5 | 588 |