Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146599.2 | NP_666810.1 | 1 | 547 | ||
GenBank mRNA | BC138389.1 | - | 1 | 547 | 8861409 | |
BC145999.1 | - | 1 | 547 | 40131302 |