Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146503.1 | NP_666714.1 | 1 | 716 | ||
GenBank mRNA | BC150723.1 | - | 1 | 794 | 9087634 |