Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146548.1 | NP_666759.1 | 1 | 415 | ||
GenBank mRNA | BC134373.1 | - | 1 | 419 | 40134856 |