Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146596.1 | NP_666807.1 | 1 | 615 | ||
GenBank mRNA | BC111815.1 | - | 1 | 614 |