Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146346.1 | NP_666458.1 | 1 | 893 | ||
GenBank mRNA | BC120574.1 | - | 1 | 973 | 40129497 | |
BC125526.1 | - | 1 | 973 | 40130041 |