Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146365.2 | NP_666477.1 | 1 | 823 | ||
GenBank mRNA | BC120867.1 | - | 1 | 823 | 40129790 |
Set Membership: |
G-protein coupled receptor |