Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146780.2 | NP_666991.1 | 1 | 339 | ||
GenBank mRNA | BC139445.1 | - | 1 | 340 | 8862467 | |
BC139446.1 | - | 1 | 340 | 8862468 |
Gene Symbol: Gm10081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162940.1 | NP_001156412.1 | 1 | 248 | ||
GenBank mRNA | BC139756.1 | - | 1 | 239 |