Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000951.2 | NP_000942.1 | 2, 3 | 245 | ||
XM_006723286.2 | XP_006723349.1 | 3, 4 | 248 | |||
GenBank mRNA | AF009243.1 | - | 1, 2 | 89 | ||
CR456846.1 | - | 1, 2 | 80 |