Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL12P38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090983.1 | - | 1 | 1945 |
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511663.2 | XP_011509965.1 | 47 | 7573 |
Gene Symbol: RPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000976.3 | NP_000967.1 | 3 | 285 | ||
GenBank mRNA | AK026526.1 | - | 1 | 258 | ||
AK057602.1 | - | 1 | 1756 | |||
BC050644.1 | - | 1 | 238 | 5108312 | ||
BC094831.1 | - | 1 | 233 | 5473344 | ||
BC105603.1 | - | 1 | 262 | 6698283 | ||
CB155565.1 | - | 3 | 285 | |||
L06505.1 | - | 3 | 238 |
Gene Symbol: RPL12P16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209471.1 | - | 1 | 268 |