Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146828.1 | NP_667039.1 | 1 | 76 | ||
GenBank mRNA | BC100677.1 | - | 1 | 75 | ||
BC101947.1 | - | 1 | 75 | |||
BC141889.1 | - | 1 | 74 |