Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKBKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193321.1 | NP_001180250.1 | 3 | 581 | ||
NM_001193322.1 | NP_001180251.1 | 4 | 700 | |||
NM_014002.3 | NP_054721.1 | 4 | 700 | |||
XM_005273356.2 | XP_005273413.1 | 4 | 696 | |||
XM_017002868.1 | XP_016858357.1 | 3 | 581 | |||
GenBank mRNA | AB016590.1 | - | 2 | 124 | ||
AF241789.1 | - | 2 | 122 | |||
AK310737.1 | - | 4 | 467 | |||
AL706097.1 | - | 4 | 491 | |||
BC105923.1 | - | 4 | 448 | 40001562 | ||
BC105924.1 | - | 4 | 448 | 40001567 | ||
BC107591.1 | - | 4 | 448 | |||
BC107812.1 | - | 3 | 329 | 40001564 | ||
D63485.1 | - | 4 | 448 | |||
DA990116.1 | - | 4 | 496 | |||
DB152295.1 | - | 3 | 203 |