Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000989.3 | NP_000980.1 | 3 | 244 | ||
GenBank mRNA | AK308890.1 | - | 3 | 204 | ||
AK312102.1 | - | 3 | 197 | |||
BC032700.2 | - | 2 | 172 | 5393199 | ||
BC095426.1 | - | 3 | 193 | 30792764 | ||
BM785413.1 | - | 3 | 244 | |||
BQ055546.1 | - | 2 | 146 | |||
L05095.1 | - | 3 | 183 | |||
X79238.1 | - | 2 | 129 |