Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146831.1 | NP_667042.1 | 1 | 596 | ||
GenBank mRNA | BC132057.1 | - | 1 | 685 | 40130334 | |
BC132059.1 | - | 1 | 685 | 40130336 |