Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010.2 | NP_001001.2 | 2, 3 | 188 | ||
GenBank mRNA | AB062123.1 | - | 2, 3 | 174 | ||
AK093634.1 | - | 1389 | ||||
AK291517.1 | - | 2, 3 | 187 | |||
AK311861.1 | - | 2, 3 | 185 | |||
BC000524.2 | - | 1, 2 | 150 | 2961137 | ||
BC009427.2 | - | 1, 2 | 141 | |||
BC013296.2 | - | 2, 3 | 171 | 3030319 | ||
BC027620.1 | - | 2, 3 | 191 | 4698843 | ||
BC035447.1 | - | 1 | 12 | |||
BC071907.1 | - | 2, 3 | 176 | 4900361 | ||
BC071908.1 | - | 2, 3 | 176 | 5089456 | ||
BC094826.1 | - | 2, 3 | 172 | 5481378 | ||
J03537.1 | - | 1, 2 | 146 | |||
M20020.1 | - | 2, 3 | 173 |
Gene Symbol: RPS6P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052613.1 | - | 1 | 531 |