Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011.3 | NP_001002.1 | 3, 4 | 261 | ||
GenBank mRNA | AF085824.1 | - | 2, 3 | 132 | ||
AK311794.1 | - | 3, 4 | 199 | |||
BC002866.2 | - | 3, 4 | 217 | 3940495 | ||
BC061901.1 | - | 3, 4 | 254 | 4442840 | ||
BC071919.1 | - | 1 | 181 | 5811776 | ||
BG615667.1 | - | 3, 4 | 262 | |||
M77233.1 | - | 2, 3 | 150 |