Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146900.2 | NP_667111.2 | 1 | 737 | ||
GenBank mRNA | BC125363.1 | - | 1 | 737 | 40129878 | |
BC131900.1 | - | 1 | 737 | 40130177 |