Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023.3 | NP_001014.1 | 3 | 330 | ||
NM_001146227.1 | NP_001139699.1 | 3 | 330 | |||
GenBank mRNA | AK130961.1 | - | 3 | 255 | ||
AK301342.1 | - | 3 | 259 | |||
AK311808.1 | - | 3 | 255 | |||
AV716603.1 | - | 259 | ||||
BC007507.2 | - | 3 | 239 | 3029762 | ||
BC087850.1 | - | 3 | 225 | 30386230 | ||
BQ064297.1 | - | 3 | 240 | |||
CB138358.1 | - | 3 | 330 | |||
L06498.1 | - | 3 | 245 |