Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147003.1 | NP_667214.1 | 1 | 725 | ||
GenBank mRNA | BC104101.2 | - | 1 | 725 | 40043110 | |
BC104102.1 | - | 1 | 725 | 40043111 |