Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147011.1 | NP_667222.1 | 1 | 886 | ||
GenBank mRNA | BC111826.1 | - | 1 | 885 |