Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147109.1 | NP_667320.1 | 1 | 709 | ||
GenBank mRNA | BC104082.1 | - | 1 | 708 | ||
BC104083.1 | - | 1 | 708 | |||
BC119411.1 | - | 1 | 709 | 40042081 | ||
BC119412.2 | - | 1 | 709 |