Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147111.1 | NP_667322.1 | 1 | 642 | ||
GenBank mRNA | AF121985.1 | - | 1 | 244 | ||
BC101949.1 | - | 1 | 641 | 40043610 | ||
BC103663.1 | - | 1 | 641 | 40043468 | ||
BC104726.1 | - | 1 | 641 | 40043611 |