Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282071.2 | NP_001269000.1 | 3 | 239 | ||
NM_147779.2 | NP_680088.1 | 3 | 239 |