Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081456.1 | NP_001074925.1 | 3 | 260 | ||
NM_148937.2 | NP_683739.2 | 3 | 411 | |||
GenBank mRNA | AK016945.1 | - | 3 | 408 | ||
AK029561.1 | - | 3 | 406 | |||
AK039149.1 | - | 3 | 238 | |||
AK140231.1 | - | 3 | 247 | |||
AY033991.1 | - | 3 | 243 | |||
BC066156.1 | - | 3 | 151 | 30099811 |