Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148945.2 | NP_683747.1 | 2 | 451 | ||
GenBank mRNA | AK079102.1 | - | 544 | |||
AK154778.1 | - | 2 | 364 | |||
AY083469.1 | - | 2 | 113 | |||
BC150156.1 | - | 2 | 116 | |||
BC150478.1 | - | 2 | 116 | |||
CJ123041.1 | - | 4 | 582 |