Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | A24059.1 | - | 3 | 320 | ||
AK313941.1 | - | 5 | 372 | |||
BC014309.2 | - | 4 | 344 | 4042766 | ||
J05125.1 | - | 4 | 356 | |||
M93285.1 | - | 4 | 344 |