Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152818.2 | NP_690031.2 | 9 | 1998 | ||
GenBank mRNA | AK007088.1 | - | 5 | 579 | ||
AK044598.1 | - | 6 | 787 | |||
BC031794.1 | - | 7 | 961 | 4501119 | ||
BC051185.1 | - | 7 | 958 | |||
BC058356.1 | - | 8 | 1141 | 5696357 | ||
BC058602.1 | - | 7 | 961 | 6467653 |