Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153056.2 | NP_694696.2 | 7, 8 | 857 | ||
GenBank mRNA | AB221687.1 | - | 3, 4 | 377 | ||
AY251540.1 | - | 7, 8 | 833 | |||
BC026403.1 | - | 7, 8 | 822 | 4209498 | ||
BC026637.1 | - | 6, 7 | 898 | |||
BC026650.1 | - | 7, 8 | 825 | 4987746 | ||
BC040759.1 | - | 7, 8 | 822 | 4209498 |