Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148926.2 | NP_683728.1 | 5 | 699 | ||
GenBank mRNA | AK082915.1 | - | 4 | 695 | ||
AK148316.1 | - | 728 | ||||
AK160675.1 | - | 6 | 622 | |||
BC024483.1 | - | 4 | 360 | |||
BC027161.1 | - | 4 | 497 | |||
BC034396.1 | - | 5 | 619 | 3707476 | ||
BC083124.1 | - | 5 | 699 | 6334368 | ||
BC100559.1 | - | 6 | 616 | 3419074 |