Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199186.1 | NP_001186115.1 | 9 | 734 | ||
NM_152808.3 | NP_690021.1 | 9 | 736 | |||
GenBank mRNA | AK029457.1 | - | 8 | 793 | ||
AK041533.1 | - | 9 | 693 | |||
AK043924.1 | - | 8 | 803 | |||
AK048648.1 | - | 9 | 721 | |||
AK088553.1 | - | 8 | 780 | |||
AK153992.1 | - | 9 | 715 | |||
AK166695.1 | - | 9 | 737 | |||
AK219076.1 | - | 2 | 117 | |||
BC013213.1 | - | 2 | 129 | |||
BC031535.1 | - | 9 | 712 | 3991609 |