Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152812.3 | NP_690025.2 | 2, 3 | 451 | ||
GenBank mRNA | AK011203.1 | - | 2, 3 | 261 | ||
AK155761.1 | - | 2, 3 | 302 | |||
AK166445.1 | - | 2, 3 | 281 | |||
AK166546.1 | - | 2, 3 | 278 | |||
AK169134.1 | - | 2, 3 | 266 | |||
AK172229.1 | - | 2, 3 | 256 | |||
AK172338.1 | - | 2, 3 | 256 | |||
BC031474.1 | - | 2, 3 | 257 | 2599116 | ||
BC087552.1 | - | 2, 3 | 248 | 6331952 | ||
CN724613.1 | - | 2, 3 | 451 |