Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025947.2 | NP_001021118.1 | 6 | 654 | ||
NM_001276340.1 | NP_001263269.1 | 6 | 672 | |||
NM_001276341.1 | NP_001263270.1 | 5 | 416 | |||
NM_152816.3 | NP_690029.2 | 7 | 693 | |||
GenBank mRNA | AB079133.1 | - | 6 | 506 | ||
AK018195.1 | - | 7 | 633 | |||
AK051443.1 | - | 6 | 583 | |||
AK080871.1 | - | 6 | 574 | |||
AK154123.1 | - | 6 | 423 | |||
BB612294.1 | - | 7 | 521 | |||
BC027538.1 | - | 6 | 574 | |||
BC040777.1 | - | 5 | 386 | 4948368 | ||
BC079635.1 | - | 6 | 503 | 6400313 | ||
BC085158.1 | - | 7 | 598 | |||
CD561569.1 | - | 5 | 380 | |||
CK634757.1 | - | 4 | 225 | |||
CN525473.1 | - | 1 | 44 | |||
CO432650.1 | - | 3 | 182 |