Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152818.2 | NP_690031.2 | 11, 12 | 2225 | ||
GenBank mRNA | AK007088.1 | - | 7, 8 | 806 | ||
AK044598.1 | - | 8, 9 | 1014 | |||
BC031794.1 | - | 9, 10 | 1188 | 4501119 | ||
BC034567.1 | - | 1, 2 | 65 | |||
BC051185.1 | - | 9, 10 | 1185 | |||
BC058356.1 | - | 10, 11 | 1368 | 5696357 | ||
BC058602.1 | - | 9, 10 | 1188 | 6467653 |