Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000954.5 | NP_000945.3 | 5 | 550 | ||
GenBank mRNA | AK075333.1 | - | 5 | 550 | ||
AK312817.1 | - | 5 | 537 | |||
AY026356.1 | - | 5 | 475 | |||
BC005939.1 | - | 5 | 484 | 4294999 | ||
BC041463.1 | - | 7 | 1758 | |||
BG037037.1 | - | 5 | 554 | |||
BM686612.1 | - | 5 | 538 | |||
BT019921.1 | - | 5 | 475 | |||
BT019922.1 | - | 5 | 475 | |||
M61900.2 | - | 5 | 540 |