Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110253.2 | NP_001103723.2 | 18 | 7294 | ||
NM_148925.3 | NP_683727.3 | 19 | 7541 | |||
GenBank mRNA | AJ428065.1 | - | 19 | 7440 | ||
AK156675.1 | - | 1 | 2173 | |||
AK162273.1 | - | 1 | 2431 |