Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pom121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148932.2 | NP_683734.2 | 13 | 4478 | ||
GenBank mRNA | AF516680.1 | - | 13 | 4439 | ||
AK154473.1 | - | 13 | 4479 | |||
AK162253.1 | - | 1 | 54 | |||
AK172982.1 | - | 12 | 3857 | |||
BC028835.1 | - | 1 | 392 | |||
BC053101.1 | - | 13 | 4454 | 6405486 |