Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000973.4 | NP_000964.1 | 4 | 491 | ||
NM_001317771.1 | NP_001304700.1 | 4 | 534 | |||
NM_001317782.1 | NP_001304711.1 | 3 | 659 | |||
NM_033301.2 | NP_150644.1 | 4 | 555 | |||
XM_017013723.1 | XP_016869212.1 | 4 | 466 | |||
GenBank mRNA | AK289459.1 | - | 4 | 442 | ||
AK301073.1 | - | 4 | 435 | |||
AK309619.1 | - | 3 | 743 | |||
BC000047.2 | - | 2 | 263 | |||
BC000077.2 | - | 4 | 426 | 3506015 | ||
BC012197.1 | - | 4 | 510 | 2905945 | ||
BC013104.1 | - | 3 | 423 | 3924874 | ||
BC030151.2 | - | 3 | 413 | |||
BC093064.1 | - | 4 | 465 | 30716964 | ||
BG480561.1 | - | 3 | 613 | |||
BI461661.1 | - | 4 | 496 | |||
BM783910.1 | - | 1 | 67 | |||
BQ929065.1 | - | 4 | 502 | |||
BT007379.1 | - | 3 | 399 | |||
CR457046.1 | - | 3 | 399 | |||
Z28407.1 | - | 4 | 442 |