Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199186.1 | NP_001186115.1 | 22 | 3016 | ||
NM_152808.3 | NP_690021.1 | 22 | 3018 | |||
GenBank mRNA | AK003942.1 | - | 1 | 620 | ||
AK041533.1 | - | 22 | 2975 | |||
AK048648.1 | - | 22 | 3003 | |||
AK153992.1 | - | 22 | 2989 | |||
AK166695.1 | - | 22 | 3019 | |||
BC013213.1 | - | 15 | 2411 | |||
BC023763.1 | - | 16 | 2253 | |||
BC031535.1 | - | 22 | 2994 | 3991609 |