Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152811.1 | NP_690024.1 | 6 | 1768 | ||
GenBank mRNA | AK004971.1 | - | 6 | 1795 | ||
AK050435.1 | - | 6 | 1796 | |||
BC027200.1 | - | 6 | 1768 | 4161686 |