Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 6, 7 | 1228 | ||
GenBank mRNA | AK046669.1 | - | 6, 7 | 1194 | ||
AK144950.1 | - | 2301 | ||||
AK164809.1 | - | 5, 6 | 1470 | |||
AK166977.1 | - | 4, 5 | 2569 | |||
AK173311.1 | - | 5, 6 | 1476 | |||
BC031392.1 | - | 6, 7 | 1172 | 4952407 |